Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxybenzeneboronic acid, 98%

Catalog No. p-7027654
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2457103 1 g
AAB2457106 5 g
AAB2457114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2457103 Supplier Thermo Scientific Chemicals Supplier No. B2457103
Only null left
Add to Cart

CAS: 107099-99-0 | C8H11BO4 | 181.982 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107099-99-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD01318181
InChI Key QOZLFNQLIKOGDR-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl boronic acid, 2,5-dimethoxybenzeneboronic acid, 2,5-dimethoxyphenylboronicacid, 2,5-dimethoxyphenyl boranediol, boronic acid, 2,5-dimethoxyphenyl, pubchem1824, acmc-2098nk, ksc174i7n, 2,5-dimethoxyphenylboronoic acid
PubChem CID 2734342
IUPAC Name (2,5-dimethoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OC)OC)(O)O

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Beilstein 2969872
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.