Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxybenzylamine 98.0+%, TCI America™
SDP

Catalog No. D40235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40235G 5 g
D402325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40235G Supplier TCI America Supplier No. D40235G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3275-95-4
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052811
InChI Key UWMCHDDHXMFKMA-UHFFFAOYSA-N
Synonym 2,5-dimethoxybenzylamine, 2,5-dimethoxyphenyl methanamine, zilain, benzenemethanamine, 2,5-dimethoxy, 1-2,5-dimethoxyphenyl methanamine, 2,5-dimethoxy-benzylamine, 2,5-dimethoxybenzenemethanamine, benzylamine, 2,5-dimethoxy, pubchem7396, acmc-1cs2n
PubChem CID 18652
IUPAC Name (2,5-dimethoxyphenyl)methanamine
SMILES COC1=CC(=C(C=C1)OC)CN

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 2735
Formula Weight 167.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethoxybenzylamine
RTECSNumber DP4430000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.