Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxyphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D144525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144525G 25 g
D14455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D144525G Supplier TCI America Supplier No. D144525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1758-25-4
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00004322
InChI Key BBZDYQUXRFATHZ-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid
PubChem CID 74469
IUPAC Name 2-(2,5-dimethoxyphenyl)acetic acid
SMILES COC1=CC(=C(C=C1)OC)CC(=O)O

Specifications

Melting Point 124°C
Color Yellow
Quantity 25 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.