Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-4-methoxy-3(2H)-furanone 97.0+%, TCI America™
SDP

Catalog No. D349925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D349925G 25 g
D34995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D349925G Supplier TCI America Supplier No. D349925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4077-47-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00209504
InChI Key SIMKGHMLPVDSJE-UHFFFAOYNA-N
PubChem CID 61325
IUPAC Name 4-methoxy-2,5-dimethyl-2,3-dihydrofuran-3-one
SMILES COC1=C(C)OC(C)C1=O

Specifications

Color Yellow
Boiling Point 63°C
Quantity 25 g
Formula Weight 142.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-4-methoxy-3(2H)-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.