Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzaldehyde, 98%

Catalog No. AAL0192406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0192406 5 g
AAL0192414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0192406 Supplier Thermo Scientific Chemicals Supplier No. L0192406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5779-94-2 | C9H10O | 134.18 g/mol

2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

Solubility
Not miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Do not expose to air. Incompatible with Air, strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5779-94-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003342
InChI Key SMUVABOERCFKRW-UHFFFAOYSA-N
PubChem CID 22015
IUPAC Name 2,5-dimethylbenzaldehyde
SMILES CC1=CC=C(C)C(C=O)=C1

Specifications

Density 0.95
Boiling Point 104°C to 106°C (14 mmHg)
Flash Point 87°C (188°F)
Odor Almond-like
Refractive Index 1.544
Quantity 5 g
Beilstein 1927303
Sensitivity Air Sensitive
Synonym benzaldehyde, 2,5-dimethyl, isoxylaldehyde, 2,5-dimethyl-benzaldehyde, acmc-20aoaf, pubchem1474, benzaldehyde,2,5-dimethyl, 2,5-dimethylbenzaldehyde
Solubility Information Not miscible in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylbenzaldehyde
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 227-303-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.