Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzoxazole 98.0+%, TCI America™
SDP

Catalog No. D346125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D346125G 25 g
D34615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D346125G Supplier TCI America Supplier No. D346125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5676-58-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005773
InChI Key XVQGFGKAPKEUFT-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzoxazole, 2,5-dimethylbenzo d oxazole, benzoxazole, 2,5-dimethyl, 2,5-dimethyl benzoxazole, pubchem8640, acmc-209lua, benzoxazole,5-dimethyl, 2,5-dimethyl-benzoxazole, benzoxazole,2,5-dimethyl, #
PubChem CID 21884
IUPAC Name 2,5-dimethyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylbenzoxazole
EINECSNumber (5)-0157
RTECSNumber DM4821600
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.