Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzylamine 97.0+%, TCI America™
SDP

Catalog No. D45225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45225G Supplier TCI America Supplier No. D45225G

missing translation for 'validMainframeMsg'

Specifications

CAS 93-48-1
Color Yellow
Molecular Formula C9H13N
Quantity 5 g
MDL Number MFCD00039780
UN Number 2735
Synonym 2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
InChI Key LUJNPFWZXIGIPS-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C)CN
IUPAC Name (2,5-dimethylphenyl)methanamine
Molecular Weight (g/mol) 135.21
PubChem CID 66735
Formula Weight 135.21
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylbenzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.