Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenol, 99+%

Catalog No. AC116540050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC116540050 5 g Glass bottle
AC116541000 100 g Glass bottle
AC116545000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC116540050 Supplier Thermo Scientific Chemicals Supplier No. 116540050
Only null left
Add to Cart

CAS: 95-87-4 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-87-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002237
InChI Key NKTOLZVEWDHZMU-UHFFFAOYSA-N
Synonym 2,5-xylenol, p-xylenol, phenol, 2,5-dimethyl, 3,6-dimethylphenol, 6-methyl-m-cresol, 3,6-xylenol, 2,5-dimethyl phenol, 2,5-dmp, 1,2,5-xylenol, 1-hydroxy-2,5-dimethylbenzene
PubChem CID 7267
IUPAC Name 2,5-dimethylphenol
SMILES CC1=CC=C(C)C(O)=C1

Specifications

Melting Point 75°C to 77°C
Color White to Cream
CAS Min % 99.0
CAS Max % 100.0
Density 0.9700g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals
Specific Gravity 0.97
Assay Percent Range 99% min. (GC)
Linear Formula (CH3)2C6H3OH
Viscosity 1.7 mPa.s (75°C)
Beilstein 06, 494
Merck Index 15, 10279
Boiling Point 212°C
Solubility Information Solubility in water: 1% (60°C). Other solubilities: soluble in naoh solution,freely soluble in alcohol,chloroform,ether and,benzene
Formula Weight 122.17
Percent Purity 99+%
Chemical Name or Material 2, 5-Dimethylphenol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-461-5
RTECSNumber ZE5775000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.