Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylterephthalic Acid 97.0+%, TCI America™
SDP

Catalog No. D220825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D220825G 25 g
D22085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D220825G Supplier TCI America Supplier No. D220825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6051-66-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00020292
InChI Key FKUJGZJNDUGCFU-UHFFFAOYSA-N
PubChem CID 299989
IUPAC Name 2,5-dimethylterephthalic acid
SMILES CC1=CC(=C(C=C1C(=O)O)C)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethylterephthalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.