Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Norbornadiene (stabilized with BHT) 97.0+%, TCI America™
SDP

Catalog No. N0346100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0346100ML 100 mL
N034625ML 25 mL
N0346500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0346100ML Supplier TCI America Supplier No. N0346100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 121-46-0
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00082301
InChI Key SJYNFBVQFBRSIB-UHFFFAOYSA-N
Synonym 2,5-norbornadiene, bicyclo 2.2.1 hepta-2,5-diene, norbornadiene, unii-w9ztq75zus, bicyclo 2.2.1 heptadiene, 8,9,10-trinorborna-2,5-diene, w9ztq75zus, dicycloheptadiene
PubChem CID 8473
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene
SMILES C1C2C=CC1C=C2

Specifications

Color Yellow
Boiling Point 90°C
Quantity 100 mL
UN Number 2251
Formula Weight 92.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Norbornadiene (stabilized with BHT)
EINECSNumber (4)-1220
RTECSNumber RB6535000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.