Learn More
Sigma Organic Chemistry 2-5-Pyrimidinyl benzald | 100mg | 640769-71-7 | MFCD02684096
Supplier: Sigma Organic Chemistry CDS005985100MG

2-5-Pyrimidinyl benzald , 100mg
About this Item:
CAS #: 640769-71-7
MDL #: MFCD02684096
Molecular Weight: 184.19
UNSPSC Code: 12352200
Chemical Formula: C11H8N2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.