Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole 98.0+%, TCI America™
SDP

Catalog No. H071925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H071925G 25 g
H0719500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H071925G Supplier TCI America Supplier No. H071925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3147-76-0
Molecular Formula C16H17N3O
Molecular Weight (g/mol) 267.332
MDL Number MFCD03844795
InChI Key WXHVQMGINBSVAY-UHFFFAOYSA-N
PubChem CID 76605
IUPAC Name 2-(benzotriazol-2-yl)-4-tert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 25 g
Formula Weight 267.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole
EINECSNumber (5)-3606
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.