Learn More
Leica Biosystems 2.5ml NCL-DYS3
Supplier:  Leica Biosystems DYS3A
Encompass
Dystrophin AntibodiesDystrophin is the 427kD protein product of the DMD gene located on the X chromosome at position Xp21.Analyte Specific Reagent. Analytical and performance characteristics are not established.
Catalog No. 50-255-2267
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.