Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene 97.0+%, TCI America™
SDP

Catalog No. B41581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41581G 1 g
B41585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41581G Supplier TCI America Supplier No. B41581G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 861398-06-3
Molecular Formula C22H28B2O4S2
Molecular Weight (g/mol) 442.202
InChI Key NTSDQKBGUDHDGQ-UHFFFAOYSA-N
PubChem CID 70976318
IUPAC Name 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[2,3-f][1]benzothiol-6-yl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC4=C(C=C(S4)B5OC(C(O5)(C)C)(C)C)C=C3S2

Specifications

Quantity 1 g
Synonym Benzo[1,2-b:4,5-b′C]dithiophene-2,6-diboronic Acid Bis(pinacol) Ester
Formula Weight 442.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b′]dithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.