Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(9H-carbazol-9-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B49611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49611G 1 g
B4961200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49611G Supplier TCI America Supplier No. B49611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 168127-49-9
Molecular Formula C29H19N3
Molecular Weight (g/mol) 409.492
MDL Number MFCD28138085
InChI Key CUQGKGMUSQKHFO-UHFFFAOYSA-N
PubChem CID 23062165
IUPAC Name 9-(6-carbazol-9-ylpyridin-2-yl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=NC(=CC=C4)N5C6=CC=CC=C6C7=CC=CC=C75

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 1 g
Formula Weight 409.49
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(9H-carbazol-9-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.