Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(hydroxymethyl)-p-cresol 98.0+%, TCI America™
SDP

Catalog No. B1525250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1525250G 250 g
B152525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1525250G Supplier TCI America Supplier No. B1525250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91-04-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004619
InChI Key KUMMBDBTERQYCG-UHFFFAOYSA-N
Synonym 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
PubChem CID 7039
IUPAC Name 2,6-bis(hydroxymethyl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)CO)O)CO

Specifications

Melting Point 126°C
Quantity 250 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(hydroxymethyl)-p-cresol
RTECSNumber CZ6440000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.