Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(p-tolyl)pyridine, 97%

Catalog No. AAB2489403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2489403 1 g
AAB2489406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2489403 Supplier Thermo Scientific Chemicals Supplier No. B2489403
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 14435-88-2 | C19H17N | 259.352 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14435-88-2
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
MDL Number MFCD00006282
InChI Key BLRYHCTZTSRBNB-UHFFFAOYSA-N
PubChem CID 84435
IUPAC Name 2,6-bis(4-methylphenyl)pyridine
SMILES CC1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=C(C=C3)C

Specifications

Melting Point 164°C to 166°C
Quantity 1 g
Synonym 2,6-bis p-tolyl pyridine, 2,6-di 4-methylphenyl pyridine, 2,6-di-p-tolylpyridine, 2,6-bis 4-methylphenyl pyridine, 2,6 di-p-tolylpyridine, 2,6-di-p-tolyl-pyridine, pyridine,2,6-di-p-tolyl, pyridine, 2,6-di-p-tolyl, 2,6-di-4-methylphenyl pyridine, pyridine, 2,6-bis 4-methylphenyl
Formula Weight 259.35
Percent Purity 97%
Chemical Name or Material 2,6-Bis(p-tolyl)pyridine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-409-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.