Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Di-tert-butyl-4-methylphenol, 99.8%

Catalog No. p-102830
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219830500 50 g Plastic bottle
AC219832500 250 g Plastic bottle
AC219830010 1 kg Plastic bottle
AC219830050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC219830500 Supplier Thermo Scientific Chemicals Supplier No. 219830500
Only null left
Add to Cart

CAS: 128-37-0 | C15H24O | 220.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.35
MDL Number MFCD00011644
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.75
CAS Max % 100.0
Melting Point 69.0°C to 71.0°C
Color White
Boiling Point 265.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 99.8% min. (GC)
Packaging Plastic bottle
Linear Formula [(CH3)3C]2C6H2(CH3)OH
Quantity 50 g
Beilstein 06,III,2073
Fieser 15,113
Merck Index 15,155
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in toluene,soluble in methanol,ethanol,isopropanol,acetone,benzene,methyl ethyl ketone,petroleum ether,,cellowolve and most other hydrocarbon solvents
Viscosity 3.47 mm2/s (0°C)
Formula Weight 220.35
Percent Purity 99.8%
Physical Form Crystals
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylphenol, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber 204-881-4
RTECSNumber GO7875000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.