Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Di(tert-butyl)pyridine, 97%, Thermo Scientific™, Thermo Scientific™

Catalog No. p-2696830
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB09883DA 1 g Amber Glass Bottle
BTB09883EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB09883DA Supplier Thermo Scientific Chemicals Supplier No. BTB09883DA
Only null left
Add to Cart

Chemical Identifiers

CAS 585-48-8
Molecular Formula C13H21N
Molecular Weight (g/mol) 191.318
MDL Number MFCD00006306
InChI Key UWKQJZCTQGMHKD-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119
PubChem CID 68510
IUPAC Name 2,6-ditert-butylpyridine
SMILES CC(C)(C)C1=NC(=CC=C1)C(C)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 0.8520g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (C5H3N[C(CH3)3]2
Quantity 1 g
Formula Weight 191.32
Percent Purity 97%
Chemical Name or Material 2, 6-Di(tert-butyl)pyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.