Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Di-tert-butylnaphthalene 98.0+%, TCI America™
SDP

Catalog No. D236925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D236925G 25 g
D23695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D236925G Supplier TCI America Supplier No. D236925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3905-64-4
Molecular Formula C18H24
Molecular Weight (g/mol) 240.39
MDL Number MFCD00004110
InChI Key TZGXZNWUOXLMFL-UHFFFAOYSA-N
PubChem CID 77509
IUPAC Name 2,6-di-tert-butylnaphthalene
SMILES CC(C)(C)C1=CC2=CC=C(C=C2C=C1)C(C)(C)C

Specifications

Melting Point 148°C
Color White
Quantity 25 g
Formula Weight 240.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.