Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Di-tert-butylpyridine

Catalog No. 2195841G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-958-41G 1 g
21-958-425G 25 g
21-958-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-958-41G Supplier Sigma Aldrich Supplier No. 2195841G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactivity of 2,6-di-tert-butylpyridine with iron(iii) tetraphenylporphyrin pi-cation radical has been examined by proton NMR spectroscopy
  • Reaction of 2,6-di-tert-butylpyridine with methyl iodide and methyl fluorosulfonate under high pressure has been reported.2,6-di-tert-butylpyridine was used as proton trapping agent to investigate the living polymerization of isobutylene
  • It was also used with cerric ammonium nitrate in the α-enolation of aldehydes leading to 1,4-dicarbonyl systems

Specifications

CAS 585-48-8
Density 0.852 g/mL (at 25°C (literature))
Boiling Point 100°C to 101°C (23 mmHg)
Molecular Formula C13H21N
Refractive Index n20/D 1.473 (literature)
Linear Formula C13H21N
MDL Number MFCD00006306
Quantity 1 g
Molecular Weight (g/mol) 191.31
Percent Purity ≥97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.