Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diaminoanthraquinone 97.0+%, TCI America™
SDP

Catalog No. D3180250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3180250G 250 g
D318025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3180250G Supplier TCI America Supplier No. D3180250G

Chemical Identifiers

CAS 131-14-6
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.246
InChI Key WQOWBWVMZPPPGX-UHFFFAOYSA-N
PubChem CID 8557
IUPAC Name 2,6-diaminoanthracene-9,10-dione
SMILES C1=CC2=C(C=C1N)C(=O)C3=C(C2=O)C=C(C=C3)N

Specifications

Melting Point 320°C
Color Red-Yellow
Quantity 250 g
Formula Weight 238.25
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminoanthraquinone
EINECSNumber (4)-0710
RTECSNumber CB6450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.