Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diaminopurine Monohydrate 97.0+%, TCI America™
SDP

Catalog No. D162525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D162525G 25 g
D16255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D162525G Supplier TCI America Supplier No. D162525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 402846-48-4
Molecular Formula C5H8N6O
Molecular Weight (g/mol) 168.16
MDL Number MFCD00213668
InChI Key UKADKJJNEZZNGH-UHFFFAOYSA-N
PubChem CID 19092948
IUPAC Name 7H-purine-2,6-diamine;hydrate
SMILES C1=NC2=C(N1)C(=NC(=N2)N)N.O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 150.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diaminopurine Monohydrate
RTECSNumber UO7524000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.