Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene 99.0+%, TCI America™
SDP

Catalog No. D48621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48621G 1 g
D4862200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48621G Supplier TCI America Supplier No. D48621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1187763-68-3
Molecular Formula C34H32Br2
Molecular Weight (g/mol) 600.438
InChI Key PVNXKVGTMVCYLK-UHFFFAOYSA-N
PubChem CID 118988970
IUPAC Name 2,6-dibromo-9,10-bis(4-tert-butylphenyl)anthracene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=C3C=C(C=CC3=C(C4=C2C=CC(=C4)Br)C5=CC=C(C=C5)C(C)(C)C)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 600.44
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-9,10-bis(4-tert-butylphenyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.