Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog No. p-7077721
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458190010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458190010 Supplier Thermo Scientific Chemicals Supplier No. 458190010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601-84-3
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00234254
InChI Key HQLOEBRPCVIFCT-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dibromo, 2,6-dibromobenzoicacid, 2,6-bis bromanyl benzoic acid
PubChem CID 4400143
IUPAC Name 2,6-dibromobenzoic acid
SMILES OC(=O)C1=C(Br)C=CC=C1Br

Specifications

Melting Point 141.0°C to 142.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 279.92
Percent Purity 95%
Chemical Name or Material 2,6-Dibromobenzoic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.