Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromoindophenol Sodium Salt, TCI America™
SDP

Catalog No. D01901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D01901G Supplier TCI America Supplier No. D01901G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5415-23-6
Molecular Formula C12H6Br2NNaO2
Molecular Weight (g/mol) 378.98
MDL Number MFCD00058958
InChI Key XFFDCGLVVOTKOS-UHFFFAOYSA-M
Synonym 2,6-Dibromophenolindophenol Sodium Salt
PubChem CID 23686434
IUPAC Name sodium 4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Br)C(=O)C(Br)=C1

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 378.98
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromoindophenol Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.