Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromoquinone-4-chloroimide 98.0+%, TCI America™
SDP

Catalog No. D020925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D020925G 25 g
D02095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D020925G Supplier TCI America Supplier No. D020925G

Chemical Identifiers

CAS 537-45-1
Molecular Formula C6H2Br2ClNO
Molecular Weight (g/mol) 299.346
MDL Number MFCD00013792
InChI Key JYWKEVKEKOTYEX-UHFFFAOYSA-N
Synonym 2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine
PubChem CID 10835
IUPAC Name 2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one
SMILES C1=C(C(=O)C(=CC1=NCl)Br)Br

Specifications

Melting Point 83°C
Quantity 25 g
Formula Weight 299.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromoquinone-4-chloroimide
RTECSNumber GU5435000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.