Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-cyanopyridine, 97%

Catalog No. AAH6348003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6348003 1 g
AAH6348006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6348003 Supplier Thermo Scientific Chemicals Supplier No. H6348003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 40381-90-6 | C6H2Cl2N2 | 172.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40381-90-6
Molecular Formula C6H2Cl2N2
Molecular Weight (g/mol) 172.996
MDL Number MFCD03411341
InChI Key LFCADBSUDWERJT-UHFFFAOYSA-N
Synonym 2,6-dichloronicotinonitrile, 2,6-dichloro-3-cyanopyridine, 3-pyridinecarbonitrile, 2,6-dichloro, 2,6-dichloro-nicotinonitrile, 2,6-dichloro-3-pyridinecarbonitrile, 2,6-dichloropyridine-3-carbonitrile, pubchem14370, acmc-20a0s7, ksc495i4d, 3-cyano-2,6-dichloropyridine
PubChem CID 12605538
IUPAC Name 2,6-dichloropyridine-3-carbonitrile
SMILES C1=CC(=NC(=C1C#N)Cl)Cl

Specifications

Melting Point 121°C to 125°C
Quantity 1 g
UN Number UN3439
Sensitivity Moisture sensitive
Formula Weight 173
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-cyanopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.