Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',6'-Dichloro-3'-fluoroacetophenone 97.0+%, TCI America™
SDP

Catalog No. D275025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D275025G 25 g
D27505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D275025G Supplier TCI America Supplier No. D275025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 290835-85-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD02093760
InChI Key VJBFZHHRVCPAPZ-UHFFFAOYSA-N
Synonym 2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
PubChem CID 2733982
IUPAC Name 1-(2,6-dichloro-3-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=C(Cl)C=CC(F)=C1Cl

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 207.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,6′-Dichloro-3′-fluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.