Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine, 95%, Thermo Scientific Chemicals

Catalog No. AAH3237403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3237403 1 g
AAH32374MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3237403 Supplier Thermo Scientific Chemicals Supplier No. H3237403
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1099597-81-5
Molecular Weight (g/mol) 271.07
MDL Number MFCD11100517
InChI Key QIULZDCQGAXJIB-UHFFFAOYSA-N
IUPAC Name 2,4-dichloro-6-(4-fluorobenzoyl)pyrimidine
SMILES FC1=CC=C(C=C1)C(=O)C1=NC(Cl)=NC(Cl)=C1

Specifications

Melting Point 95°C to 99°C
Molecular Formula C11H5Cl2FN2O
Quantity 1 g
Formula Weight 271.08
Percent Purity 95%
Chemical Name or Material 2,6-Dichloro-4-(4-fluorobenzoyl)pyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.