Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-fluoroaniline 97.0+%, TCI America™
SDP

Catalog No. D36251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36251G 1 g
D36255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36251G Supplier TCI America Supplier No. D36251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 344-19-4
Molecular Formula C6H4Cl2FN
Molecular Weight (g/mol) 180.003
MDL Number MFCD00142845
InChI Key YAUYKCFMKMZTEX-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-fluorobenzenamine, 2,6-dichloro-4-fluoro-aniline, 2',6'-dichloro-4'-fluoroaniline, benzenamine, 2,6-dichloro-4-fluoro, 2,6-dichloro-4-fluorophenylamine, pubchem3451, acmc-1ad5c, ksc572g5t, 2,6,-dichloro-4-fluoroaniline, 2,6 dichloro-4-fluorophenylamine
PubChem CID 2774008
IUPAC Name 2,6-dichloro-4-fluoroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)F

Specifications

Melting Point 56°C
Quantity 1 g
UN Number 2811
Formula Weight 180.00
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.