Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. D380225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D380225G 25 g
D38025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D380225G Supplier TCI America Supplier No. D380225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99479-66-0
Molecular Formula C7H4Cl2F3NO
Molecular Weight (g/mol) 246.01
MDL Number MFCD00190128
InChI Key FKISQWQHZULEEG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethoxy aniline, benzenamine, 2,6-dichloro-4-trifluoromethoxy, 2,6-dichloro-4-trifluoromethoxy phenylamine, pubchem8461, acmc-209scs, 3.5-dichloro-4-amino trifluoromethoxybenzene, 2,6-dichloro-4-trifluoromethoxy-phenylamine, 3,5-dichloro-4-amino trifluoromethoxybenzene, 3,5-dichloro-4-aminotrifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy benzenamine
PubChem CID 688247
IUPAC Name 2,6-dichloro-4-(trifluoromethoxy)aniline
SMILES NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl

Specifications

Melting Point 33°C
Quantity 25 g
Formula Weight 246.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.