Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzene-1-sulfonyl chloride, ≥95%, Thermo Scientific™

Catalog No. TL00344DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00344DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00344DA Supplier Thermo Scientific Chemicals Supplier No. TL00344DA
Add to Cart

Chemical Identifiers

CAS 6579-54-0
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052311
InChI Key WGGKQIKICKLWGN-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
PubChem CID 522183
IUPAC Name 2,6-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3SO2Cl
Quantity 1 g
Formula Weight 245.51
Percent Purity 95%
Chemical Name or Material 2, 6-Dichlorobenzene-1-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.