Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D033925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D033925G 25 g
D03395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. D033925G Supplier TCI America Supplier No. D033925G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-30-6
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002418
InChI Key MRUDNSFOFOQZDA-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dichloro, 2,6-dichloro-benzoic acid, unii-634ri764qt, 2,6 dichlorobenzoic acid, 2,6-dichlorobenzoicacid, 2,6-dichloro-benzoicacid, ccris 8610, pubchem20028, acmc-1auop, 2.6-dichlorobenzoic acid
PubChem CID 5758
ChEBI CHEBI:48623
IUPAC Name 2,6-dichlorobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzoic Acid
EINECSNumber (3)-1410&(3)-1424
RTECSNumber DG7000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.