Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzonitrile 99.0+%, TCI America™
SDP

Catalog No. D1137500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1137500G 500 g
D113725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1137500G Supplier TCI America Supplier No. D1137500G

Chemical Identifiers

CAS 1194-65-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00001781
InChI Key YOYAIZYFCNQIRF-UHFFFAOYSA-N
Synonym dichlobenil, dichlobanil, casoron, prefix d, benzonitrile, 2,6-dichloro, casaron, du-sprex, decabane, surfassol, casoron 133
PubChem CID 3031
ChEBI CHEBI:943
IUPAC Name 2,6-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)Cl

Specifications

Melting Point 145°C
Color White
Quantity 500 g
UN Number 3077
Formula Weight 172.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzonitrile
EINECSNumber (3)-4103
RTECSNumber DI3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.