Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzothiazole 98.0+%, TCI America™
SDP

Catalog No. D38011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38011G 1 g
D38015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38011G Supplier TCI America Supplier No. D38011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3622-23-9
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
MDL Number MFCD00044101
InChI Key QDZGJGWDGLHVNK-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzothiazole, 2,6-dichlorobenzo d thiazole, benzothiazole, 2,6-dichloro, 2,6-dichloro-benzothiazole, 2,6-dichloro benzothiazole, pubchem17324, acmc-1adss, ksc490o9j
PubChem CID 77176
IUPAC Name 2,6-dichloro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1Cl)SC(=N2)Cl

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.