Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorocinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. D404825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D404825G 25 g
D40485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D404825G Supplier TCI America Supplier No. D404825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5345-89-1
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.05
MDL Number MFCD00004374
InChI Key OIPVGRCXMFBNAN-SNAWJCMRSA-N
Synonym 2,6-dichlorocinnamic acid, 3-2,6-dichlorophenyl acrylic acid, e-3-2,6-dichlorophenyl acrylic acid, 2e-3-2,6-dichlorophenyl prop-2-enoic acid, 2e-3-2,6-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,6-dichlorophenyl, 3-2,6-dichlorophenyl-2-propenoic acid, cinnamic acid, 2,6-dichloro-6ci,7ci, 3-2,6-dichlorophenyl propenoic acid, 2,6-dichlorocinnamic acid, predominantly trans
PubChem CID 731762
IUPAC Name (2E)-3-(2,6-dichlorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 25 g
Formula Weight 217.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorocinnamic Acid
RTECSNumber UD3333800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.