Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. D3778250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3778250G 250 g
D377825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3778250G Supplier TCI America Supplier No. D3778250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15307-93-4
Molecular Formula C12H9Cl2N
Molecular Weight (g/mol) 238.11
MDL Number MFCD00269648
InChI Key HDUUZPLYVVQTKN-UHFFFAOYSA-N
Synonym 2,6-dichlorodiphenylamine, n-phenyl-2,6-dichloroaniline, benzenamine, 2,6-dichloro-n-phenyl, 2,6-dichlorophenyl phenylamine, dichlorophenylaniline, 2,6-dichlorophenylaniline, diphenylamine,2,6-dichloro, n-phenyl 2,6-dichloroaniline, n-2,6-dichlorophenyl aniline, #
PubChem CID 84867
IUPAC Name 2,6-dichloro-N-phenylaniline
SMILES ClC1=CC=CC(Cl)=C1NC1=CC=CC=C1

Specifications

Melting Point 51°C
Quantity 250 g
Formula Weight 238.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorodiphenylamine
EINECSNumber (3)-0061
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.