Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ 2,6-Dichloroindophenol Sodium Salt
SCI_ED

Catalog No. p-4416712 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
0.036 g
5 g
Packaging:
Powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25300A 5 g Powder
S25300B 0.036 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25300A
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2, 6-Dichlorophenol Indophenol Sodium Salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Linear Formula NaOC6H4N·C6H2(Cl2)·O
Formula Weight 290.08
Packaging Powder
Color Green
Quantity 5 g
Chemical Name or Material 2, 6-Dichloroindophenol Sodium Salt
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.