Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D33451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33451G 1 g
D33455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33451G Supplier TCI America Supplier No. D33451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5398-44-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00234147
InChI Key SQSYNRCXIZHKAI-UHFFFAOYSA-N
Synonym 2,6-dichloroisonicotinic acid, 2,6-dichloro-isonicotinic acid, 2,6-dichloro-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2,6-dichloro, 2,6-dichloroisonicotinicacid, 2,6-dichloro-4-pyridinyl carboxylic acid, pyridine-4-carboxylic acid, 2,6-dichloro, 2,6-dichlorisonicotins, enamine_005327, zlchem 403
PubChem CID 94830
ChEBI CHEBI:73179
IUPAC Name 2,6-dichloropyridine-4-carboxylic acid
SMILES OC(=O)C1=CC(Cl)=NC(Cl)=C1

Specifications

Melting Point 211°C
Quantity 1 g
Formula Weight 192.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloroisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.