Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropurine 97.0+%, TCI America™
SDP

Catalog No. D247025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D247025G 25 g
D24701G 1 g
D24705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D247025G Supplier TCI America Supplier No. D247025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5451-40-1
Molecular Formula C5H2Cl2N4
Molecular Weight (g/mol) 189.00
MDL Number MFCD00077725
InChI Key RMFWVOLULURGJI-UHFFFAOYSA-N
Synonym 2,6-dichloropurine, 2,6-dichloro-9h-purine, 2,6-dichloro-1h-purine, 9h-purine, 2,6-dichloro, 2,6-dichloro purine, 2,6 dichloropurine, 2,6-dichioropurine, purine,6-dichloro, zlchem 226, 2,6-dichloro-purine
PubChem CID 5324412
IUPAC Name 2,6-dichloro-7H-purine
SMILES ClC1=NC(Cl)=C2NC=NC2=N1

Specifications

Quantity 25 g
Formula Weight 189.00
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloropurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.