Learn More
Sigma Aldrich 2,6-Dichloroquinoline-3-methanol
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2-heptanol is an aliphatic secondary alcohol
- It can undergo selective oxidation to form the corresponding ketone in the presence of a novel pd catalyst supported on amphiphilic carbon nanotubes
- The distribution coefficient of 2-heptanol between micelles and water indicates that it is solubilized both at the surface and in the interior of the micelles.2-heptanol in combination with dimethyl sulfoxide form a porogenic mixture that may be used produce macroporous poly (acrylamide-co-n,nα-methylenebisacrylamide) monolithics via copolymerization
Specifications
Specifications
| CAS | 1017429-35-4 |
| Molecular Formula | C10H7Cl2NO |
| Linear Formula | C10H7Cl2NO |
| MDL Number | MFCD09997988 |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 228.07 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.