Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diethylaniline 98.0+%, TCI America™
SDP

Catalog No. D0476100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0476100ML 100 mL
D047625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0476100ML Supplier TCI America Supplier No. D0476100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 579-66-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00007753
InChI Key FOYHNROGBXVLLX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine
PubChem CID 11369
ChEBI CHEBI:27880
IUPAC Name 2,6-diethylaniline
SMILES CCC1=C(C(=CC=C1)CC)N

Specifications

Boiling Point 216°C
Quantity 100 mL
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Diethylaniline
EINECSNumber (3)-0129
RTECSNumber BX3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.