Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diethylphenyl isocyanate, 98+%

Catalog No. p-7045613
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1133903 1 g
AAL1133906 5 g
AAL1133914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1133903 Supplier Thermo Scientific Chemicals Supplier No. L1133903
Only null left
Add to Cart

CAS: 20458-99-5 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20458-99-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00019912
InChI Key AUSRFKVPUCQCCN-UHFFFAOYSA-N
Synonym 2,6-diethylphenyl isocyanate, 2,6-diethylphenylisocyanate, 1,3-diethyl-2-isocyanato-benzene, benzene, 1,3-diethyl-2-isocyanato, acmc-1co7s, 2,6-diethylbenzenisocyanate, benzene,1,3-diethyl-2-isocyanato
PubChem CID 5153183
IUPAC Name 1,3-diethyl-2-isocyanatobenzene
SMILES CCC1=CC=CC(CC)=C1N=C=O

Specifications

Density 1.014
Boiling Point 114°C to 115°C (1 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.526
Quantity 1 g
UN Number UN2206
Beilstein 2641462
Sensitivity Moisture sensitive
Formula Weight 175.23
Percent Purity ≥98%
Chemical Name or Material 2,6-Diethylphenyl isocyanate
Show More Show Less
Hazard Category H301-H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.