Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. D51421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51421G 1 g
D5142200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51421G Supplier TCI America Supplier No. D51421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1072945-00-6
Molecular Formula C11H14BF2NO2
Molecular Weight (g/mol) 241.04
MDL Number MFCD09037475
InChI Key CLDFZDCZPCIBKO-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-difluoropyridine-3-boronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid pinacol ester, 2,6-difluoropyridin-3-ylboronic acid, pinacol ester, 2,6-difluoropyridin-3-yl boronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl pyridine, 2,6-difluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-2,6-difluoro-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 44755153
IUPAC Name 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(F)N=C(F)C=C1

Specifications

Melting Point 57°C
Color White
Quantity 1 g
Formula Weight 241.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.