Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoroaniline, 97%

Catalog No. AC188270250
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188270250 25 mL Glass bottle
AC188270050 5 mL Glass bottle
AC188271000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC188270250 Supplier Thermo Scientific Chemicals Supplier No. 188270250
Add to Cart

CAS: 5509-65-9 | C6H5F2N | 129.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5509-65-9
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007655
InChI Key ODUZJBKKYBQIBX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-difluoro, 2,6-difluoro-phenylamine, 2,6-difluoro aniline, 2,6-difluorobenzenamine, 2,6-difluoranilin, 2,6 difluoroaniline, 2,6-diflouroaniline, pubchem4034, 2,6-difuluoroaniline
PubChem CID 79647
IUPAC Name 2,6-difluoroaniline
SMILES NC1=C(F)C=CC=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.1990g/mL
Boiling Point 51°C to 52°C (15.0 mmHg)
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5070 to 1.509
Linear Formula F2C6H3NH2
Quantity 25 mL
Beilstein 03, 727H
Specific Gravity 1.199
Solubility Information Solubility in water: immiscible
Formula Weight 129.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Difluoroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or
EINECSNumber 226-853-
Recommended Storage Product darkens on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.