Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D294025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D294025G 25 g
D29401G 1 g
D29405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D294025G Supplier TCI America Supplier No. D294025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60230-36-6
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00085004
InChI Key QXWAUQMMMIMLTO-UHFFFAOYSA-N
Synonym 2,6-difluorobenzene-1-sulfonyl chloride, 2,6-difluorobenzenesulfonylchloride, 2,6-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,6-difluoro, 2,6-difluoro-benzenesulfonyl chloride, 2,6-difluorobenzene sulfonyl chloride, 2,6-difluorophenyl chlorosulfone, pubchem1957, acmc-209mil, ksc352q0t
PubChem CID 2734271
IUPAC Name 2,6-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)F)S(=O)(=O)Cl)F

Specifications

Color Yellow
Boiling Point 129°C
Quantity 25 g
UN Number 3265
Formula Weight 212.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.