Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. D451025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D451025G 25 g
D45105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D451025G Supplier TCI America Supplier No. D451025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 654-01-3
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009973
InChI Key GVAYBGQTAADLJS-UHFFFAOYSA-N
Synonym 2,6-difluorophenylacetonitrile, 2-2,6-difluorophenyl acetonitrile, 2,6-difluorobenzyl cyanide, 2,6-difluorophenyl acetonitrile, 2,6-difluorobenzeneacetonitrile, benzeneacetonitrile, 2,6-difluoro, 2-2,6-difluorophenyl ethanenitrile, pubchem4152, acmc-1axnw, n-dodecyl-beta-maltoside
PubChem CID 69559
IUPAC Name 2-(2,6-difluorophenyl)acetonitrile
SMILES FC1=CC=CC(F)=C1CC#N

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 g
UN Number 1992
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.