Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxyanthraquinone 94%, Thermo Scientific™

Catalog No. AC104890250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104890250 25 g
AC104891000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104890250 Supplier Thermo Scientific Chemicals Supplier No. 104890250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-60-6
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001228
InChI Key APAJFZPFBHMFQR-UHFFFAOYSA-N
Synonym 2,6-dihydroxyanthraquinone, anthraflavic acid, anthraflavin, 2,6-dihydroxyanthra-9,10-quinone, 9,10-anthracenedione, 2,6-dihydroxy, az-f, anthrafravic acid, anthraquinone, 2,6-dihydroxy, 2,6-dihydroxy-9,10-anthraquinone, 2,6-dihydroxy-anthraquinone
PubChem CID 6776
ChEBI CHEBI:34250
IUPAC Name 2,6-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C=C1O)C(=O)C3=C(C2=O)C=C(C=C3)O

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 94%
Quantity 25 g
Beilstein 08, 463
Formula Weight 240.21
Percent Purity ≥92%
Physical Form Powder
Chemical Name or Material 2, 6-Dihydroxyanthraquinone, 94%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-544-3
RTECSNumber CB6675000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.