Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D1959250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1959250G 250 g
D195925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1959250G Supplier TCI America Supplier No. D1959250G

Chemical Identifiers

CAS 1466-76-8
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002437
InChI Key MBIZFBDREVRUHY-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-dimethoxy, 2,6-dimethoxy-benzoic acid, unii-g6b3p1lo43, 2,6-dimethoxybenzoicacid, pubchem19743, acmc-1bwjj, intermediates-zcf02125, benzoic acid,6-dimethoxy, dsstox_cid_26999, dsstox_rid_82043
PubChem CID 15109
IUPAC Name 2,6-dimethoxybenzoic acid
SMILES COC1=CC=CC(OC)=C1C(O)=O

Specifications

Melting Point 185°C
Color White
Quantity 250 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.